siRNA Details
| Gene Symbol: 1700013F07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029314.1 | NP_083590.1 | 4 | 432 | ||
| GenBank mRNA | AK005946.1 | - | 4 | 451 | ||
| BC049763.1 | - | 4 | 470 | |||
| BC100493.1 | - | 465 | ||||
| BC116222.1 | - | 4 | 433 | |||
| BC116223.1 | - | 4 | 432 | 40091335 | ||
 Back To Top
 Back To TopMore Information
Other Species Targeted
 This siRNA also targets the following transcripts in Rat: NM_001134578, XM_008761408.
					            	This siRNA also targets the following transcripts in Rat: NM_001134578, XM_008761408.Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 1700013F07Rik
1700013F07Rik Genomic Map
Genomic Map Back To Top
 Back To Top