siRNA Details
| Gene Symbol: 1700013H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200013.1 | NP_001186942.1 | 4, 5 | 516 | ||
| GenBank mRNA | AK005953.1 | - | 4, 5 | 499 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700013H16Rik| Gene Symbol: 1700013H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200013.1 | NP_001186942.1 | 4, 5 | 516 | ||
| GenBank mRNA | AK005953.1 | - | 4, 5 | 499 | ||