siRNA Details
Gene Symbol: 1700013H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200013.1 | NP_001186942.1 | 7 | 793 | ||
GenBank mRNA | AK005953.1 | - | 7 | 776 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700013H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200013.1 | NP_001186942.1 | 7 | 793 | ||
GenBank mRNA | AK005953.1 | - | 7 | 776 |