siRNA Details
| Gene Symbol: ACPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099.4 | NP_001090.2 | 4 | 430 | ||
| NM_001134194.1 | NP_001127666.1 | 4 | 430 | |||
| NM_001292037.1 | NP_001278966.1 | 4 | 430 | |||
| XM_011512946.1 | XP_011511248.1 | 4 | 394 | |||
| XM_011512947.2 | XP_011511249.1 | 4 | 394 | |||
| GenBank mRNA | AB102888.1 | - | 4 | 340 | ||
| AK300540.1 | - | 4 | 391 | |||
| BC007460.1 | - | 4 | 390 | 3951204 | ||
| BC008493.1 | - | 4 | 387 | 4272289 | ||
| BC016344.1 | - | 4 | 394 | 3950907 | ||
| BC017877.1 | - | 4 | 380 | |||
| DC383453.1 | - | 4 | 430 | |||
| M34840.1 | - | 4 | 354 | |||
| X52174.1 | - | 4 | 363 | |||
| X53605.1 | - | 4 | 376 | |||

ACPP
Genomic Map