siRNA Details
| Gene Symbol: Lyrm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029327.3 | NP_083603.2 | 3 | 156 | ||
| GenBank mRNA | AK006294.1 | - | 3 | 482 | ||
| BC139403.1 | - | 3 | 287 | 8862425 | ||
| BC139406.1 | - | 3 | 287 | 8862428 | ||
| BY776521.1 | - | 3 | 156 | |||

Lyrm7
Genomic Map