siRNA Details
| Gene Symbol: 1700019G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029331.3 | NP_083607.3 | 4 | 1161 | ||
| GenBank mRNA | BC029200.1 | - | 1107 | 3501234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700019G17Rik| Gene Symbol: 1700019G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029331.3 | NP_083607.3 | 4 | 1161 | ||
| GenBank mRNA | BC029200.1 | - | 1107 | 3501234 | ||