siRNA Details
| Gene Symbol: Zc3h14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008506.2 | NP_001008506.2 | 2, 3 | 231 | ||
| NM_001160107.1 | NP_001153579.1 | 2, 3 | 231 | |||
| NM_029334.2 | NP_083610.2 | 2, 3 | 231 | |||
| GenBank mRNA | AK048046.1 | - | 2, 3 | 197 | ||
| AK160937.1 | - | 2, 3 | 217 | |||
| BC024824.1 | - | 2, 3 | 201 | 5355748 | ||
| BC034760.1 | - | 2, 3 | 164 | |||

Zc3h14
Genomic Map