siRNA Details
| Gene Symbol: Ccdc101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029339.3 | NP_083615.3 | 9 | 843 | ||
| GenBank mRNA | AK006287.1 | - | 9 | 753 | ||
| AK139102.1 | - | 9 | 844 | |||
| AK169898.1 | - | 9 | 839 | |||
| BC026784.1 | - | 9 | 781 | 4206090 | ||

Ccdc101
Genomic Map