siRNA Details
| Gene Symbol: Spata9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029343.3 | NP_083619.1 | 5 | 632 | ||
| GenBank mRNA | AK006554.1 | - | 5 | 633 | ||
| BC048558.1 | - | 5 | 612 | 6703288 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata9| Gene Symbol: Spata9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029343.3 | NP_083619.1 | 5 | 632 | ||
| GenBank mRNA | AK006554.1 | - | 5 | 633 | ||
| BC048558.1 | - | 5 | 612 | 6703288 | ||