siRNA Details
| Gene Symbol: 2310007L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029345.1 | NP_083621.1 | 4 | 342 | ||
| GenBank mRNA | AK009213.1 | - | 5 | 665 | ||
| AK153786.1 | - | 4 | 343 | |||
| BC116233.1 | - | 3 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310007L24Rik| Gene Symbol: 2310007L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029345.1 | NP_083621.1 | 4 | 342 | ||
| GenBank mRNA | AK009213.1 | - | 5 | 665 | ||
| AK153786.1 | - | 4 | 343 | |||
| BC116233.1 | - | 3 | 291 | |||