siRNA Details
| Gene Symbol: Zbtb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029348.2 | NP_083624.2 | 3 | 1158 | ||
| GenBank mRNA | AK147212.1 | - | 3 | 1112 | ||
| AK163593.1 | - | 3 | 1221 | |||
| AK173196.1 | - | 1 | 912 | |||
| BC112421.1 | - | 2 | 954 | |||
| BC113187.1 | - | 2 | 954 | |||

Zbtb4
Genomic Map