siRNA Details
| Gene Symbol: Yipf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023784.5 | NP_076273.1 | 3 | 303 | ||
| GenBank mRNA | AF217188.1 | - | 2 | 215 | ||
| AK009405.1 | - | 3 | 254 | |||
| BC089576.1 | - | 3 | 288 | 6758950 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yipf7| Gene Symbol: Yipf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023784.5 | NP_076273.1 | 3 | 303 | ||
| GenBank mRNA | AF217188.1 | - | 2 | 215 | ||
| AK009405.1 | - | 3 | 254 | |||
| BC089576.1 | - | 3 | 288 | 6758950 | ||