siRNA Details
| Gene Symbol: NDUFB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004146.5 | NP_004137.2 | 2 | 216 | ||
| XM_011528039.2 | XP_011526341.1 | 2 | 808 | |||
| GenBank mRNA | AF112200.1 | - | 2 | 202 | ||
| AF217091.1 | - | 2 | 161 | |||
| BC002595.2 | - | 2 | 201 | 3140806 | ||
| BU565399.1 | - | 217 | ||||
| CB124371.1 | - | 2 | 216 | |||
| CR450354.1 | - | 2 | 139 | |||

NDUFB7
Genomic Map