siRNA Details
| Gene Symbol: Krtap9-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029351.2 | NP_083627.2 | 1 | 514 | ||
| GenBank mRNA | AK009730.1 | - | 1 | 516 | ||
| AK131927.1 | - | 1 | 515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap9-3| Gene Symbol: Krtap9-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029351.2 | NP_083627.2 | 1 | 514 | ||
| GenBank mRNA | AK009730.1 | - | 1 | 516 | ||
| AK131927.1 | - | 1 | 515 | |||