siRNA Details
| Gene Symbol: Ndufaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127346.1 | NP_001120818.1 | 2, 3 | 281 | ||
| GenBank mRNA | AB183434.1 | - | 2, 3 | 225 | ||
| AK007894.1 | - | 2, 3 | 272 | |||
| AK166492.1 | - | 2, 3 | 248 | |||
| AY262012.1 | - | 2, 3 | 253 | |||
| BC048512.1 | - | 2, 3 | 240 | |||
| BC116975.1 | - | 2, 3 | 247 | 40126294 | ||
| BC116977.1 | - | 2, 3 | 247 | 40126296 | ||
| BY251841.1 | - | 2, 3 | 282 | |||

Ndufaf2
Genomic Map