siRNA Details
| Gene Symbol: Kdm5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152895.2 | NP_690855.2 | 2 | 391 | ||
| GenBank mRNA | AK008738.1 | - | 2 | 344 | ||
| AK041304.1 | - | 2 | 398 | |||
| AK163481.1 | - | 2 | 363 | |||
| AK220451.1 | - | 2 | 377 | |||
| AY082429.1 | - | 2 | 374 | |||
| AY082430.1 | - | 2 | 374 | |||
| BC048180.1 | - | 2 | 374 | 6826125 | ||
| BC057318.1 | - | 2 | 374 | 6826125 | ||

Kdm5b
Genomic Map