siRNA Details
| Gene Symbol: Wnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290311.1 | NP_001277240.1 | 16, 17 | 4110 | ||
| NM_001290313.1 | NP_001277242.1 | 16, 17 | 4110 | |||
| NM_029361.4 | NP_083637.2 | 16, 17 | 4110 | |||
| GenBank mRNA | AK129439.1 | - | 5, 6 | 967 | ||
| AK147550.1 | - | 16, 17 | 3797 | |||
| BC046464.1 | - | 9, 10 | 1909 | |||
| BC060187.1 | - | 5, 6 | 1141 | |||

Wnk2
Genomic Map