siRNA Details
| Gene Symbol: Smim15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048250.2 | NP_001041715.1 | 2 | 359 | ||
| GenBank mRNA | AK088529.1 | - | 3 | 375 | ||
| AK139507.1 | - | 2 | 360 | |||
| BC004049.1 | - | 2 | 317 | 3497350 | ||
| BC069965.1 | - | 3 | 399 | |||
| BC079864.1 | - | 2 | 350 | |||

Smim15
Genomic Map