siRNA Details
| Gene Symbol: Ssmem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027073.1 | NP_081349.1 | 3 | 832 | ||
| NM_029373.4 | NP_083649.1 | 3 | 646 | |||
| GenBank mRNA | AK006038.1 | - | 3 | 833 | ||
| AK006332.1 | - | 3 | 632 | |||
| BC049755.1 | - | 3 | 781 | 6773255 | ||
| BU938696.1 | - | 3 | 649 | |||

Ssmem1
Genomic Map