siRNA Details
| Gene Symbol: Fam65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081241.2 | NP_001074710.2 | 13 | 1964 | ||
| GenBank mRNA | AK052316.1 | - | 13 | 1913 | ||
| AK122573.1 | - | 13 | 1938 | |||
| AK180976.1 | - | 1 | 220 | |||
| BC030451.1 | - | 3 | 1027 | 5400266 | ||
| BC079880.1 | - | 13 | 1878 | 6852251 | ||

Fam65a
Genomic Map