siRNA Details
Gene Symbol: Higd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080846.1 | NP_543122.1 | 3 | 708 | ||
GenBank mRNA | AK009942.1 | - | 3 | 549 | ||
BC005568.1 | - | 3 | 708 | 3967350 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Higd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080846.1 | NP_543122.1 | 3 | 708 | ||
GenBank mRNA | AK009942.1 | - | 3 | 549 | ||
BC005568.1 | - | 3 | 708 | 3967350 |