siRNA Details
| Gene Symbol: Krt24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029393.1 | NP_083669.1 | 1 | 536 | ||
| GenBank mRNA | AK010165.1 | - | 1 | 561 | ||
| AK053678.1 | - | 1 | 282 | |||
| BC129847.1 | - | 1 | 536 | 40107361 | ||
| BC129848.1 | - | 1 | 536 | 40107363 | ||
| GU046702.1 | - | 1 | 505 | |||

Krt24
Genomic Map