siRNA Details
Gene Symbol: 4932414N04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183113.3 | NP_898936.1 | 27 | 3147 | ||
GenBank mRNA | AK016523.1 | - | 27 | 3148 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4932414N04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183113.3 | NP_898936.1 | 27 | 3147 | ||
GenBank mRNA | AK016523.1 | - | 27 | 3148 |