siRNA Details
| Gene Symbol: Morc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193309.1 | NP_001180238.1 | 12 | 1700 | ||
| NM_029413.4 | NP_083689.2 | 12 | 1700 | |||
| GenBank mRNA | AK017472.1 | - | 12 | 1669 | ||
| AK032807.1 | - | 12 | 1701 | |||
| BC036164.1 | - | 6 | 587 | |||
| BC039283.1 | - | 5 | 412 | |||
| BC098483.1 | - | 12 | 1655 | 30022418 | ||
| BC132153.1 | - | 12 | 1554 | 40130430 | ||
| BC132497.1 | - | 12 | 1554 | 40130774 | ||

Morc4
Genomic Map