siRNA Details
| Gene Symbol: Morc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193309.1 | NP_001180238.1 | 8 | 1280 | ||
| NM_029413.4 | NP_083689.2 | 8 | 1280 | |||
| GenBank mRNA | AK017472.1 | - | 8 | 1249 | ||
| AK032807.1 | - | 8 | 1281 | |||
| BC036164.1 | - | 2 | 167 | |||
| BC098483.1 | - | 8 | 1235 | 30022418 | ||
| BC132153.1 | - | 8 | 1134 | 40130430 | ||
| BC132497.1 | - | 8 | 1134 | 40130774 | ||

Morc4
Genomic Map