siRNA Details
| Gene Symbol: Apol7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164640.1 | NP_001158112.1 | 5 | 691 | ||
| NM_029419.2 | NP_083695.2 | 6 | 791 | |||
| GenBank mRNA | AK018646.1 | - | 6 | 793 | ||
| AK085956.1 | - | 5 | 668 | |||
| BC139137.1 | - | 5 | 570 | 8862159 | ||
| BC139138.1 | - | 5 | 570 | 8862160 | ||

Apol7a
Genomic Map