siRNA Details
Gene Symbol: Lppr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014763.1 | - | 3 | 499 | ||
AK032034.1 | - | 3 | 865 | |||
AK043629.1 | - | 3 | 489 | |||
AK076176.1 | - | 3 | 630 | |||
AK082692.1 | - | 3 | 761 | |||
AY512657.1 | - | 3 | 412 | |||
BC031879.1 | - | 3 | 729 | 4527789 |

More Information
Other Species Targeted

Panther Classification:
