siRNA Details
| Gene Symbol: Lca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027448.2 | NP_081724.1 | 2, 3 | 556 | ||
| NM_029434.3 | NP_083710.2 | 2, 3 | 452 | |||
| GenBank mRNA | AK015260.1 | - | 2, 3 | 424 | ||
| AK047956.1 | - | 2, 3 | 419 | |||
| BC049101.1 | - | 2, 3 | 484 | 5702365 | ||
| BC052060.1 | - | 2, 3 | 484 | 5702365 | ||
| CK639693.1 | - | 1, 2 | 293 | |||

Lca5
Genomic Map