siRNA Details
| Gene Symbol: Cdyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029441.3 | NP_083717.1 | 2 | 364 | ||
| GenBank mRNA | AK015452.1 | - | 2 | 218 | ||
| AK142084.1 | - | 2 | 254 | |||
| AY273799.1 | - | 2 | 213 | |||
| BC058956.1 | - | 2 | 364 | 5690642 | ||

Cdyl2
Genomic Map