siRNA Details
| Gene Symbol: Cdyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029441.3 | NP_083717.1 | 3 | 903 | ||
| GenBank mRNA | AK006509.1 | - | 2 | 198 | ||
| AK015452.1 | - | 3 | 757 | |||
| AK142084.1 | - | 3 | 793 | |||
| AY273799.1 | - | 3 | 752 | |||
| BC058956.1 | - | 3 | 903 | 5690642 | ||

Cdyl2
Genomic Map