siRNA Details
| Gene Symbol: Nln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029447.2 | NP_083723.1 | 4 | 655 | ||
| GenBank mRNA | AK079090.1 | - | 4 | 656 | ||
| AK138399.1 | - | 4 | 628 | |||
| AK146364.1 | - | 4 | 604 | |||
| AK170508.1 | - | 4 | 656 | |||
| AK171432.1 | - | 4 | 656 | |||
| BC016224.1 | - | 4 | 628 | 4486941 | ||

Nln
Genomic Map