siRNA Details
| Gene Symbol: Saxo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081096.1 | NP_001074565.1 | 2 | 432 | ||
| GenBank mRNA | AK015673.1 | - | 2 | 404 | ||
| AK049262.1 | - | 2 | 256 | |||
| AK076898.1 | - | 2 | 456 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Saxo1| Gene Symbol: Saxo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081096.1 | NP_001074565.1 | 2 | 432 | ||
| GenBank mRNA | AK015673.1 | - | 2 | 404 | ||
| AK049262.1 | - | 2 | 256 | |||
| AK076898.1 | - | 2 | 456 | |||