siRNA Details
| Gene Symbol: Ispd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289502.1 | NP_001276431.1 | 4, 5 | 1011 | ||
| NM_001289503.1 | NP_001276432.1 | 5, 6 | 743 | |||
| NM_178629.6 | NP_848744.2 | 5, 6 | 1161 | |||
| GenBank mRNA | AK016317.1 | - | 5, 6 | 710 | ||
| AK039004.1 | - | 5, 6 | 1029 | |||
| AK045940.1 | - | 5, 6 | 1163 | |||
| AK051322.1 | - | 4, 5 | 706 | |||
| BC024061.1 | - | 4, 5 | 443 | |||
| BC050993.1 | - | 5, 6 | 999 | |||
| BC086665.1 | - | 5, 6 | 1092 | 30612014 | ||
| BC118038.1 | - | 5, 6 | 929 | 40093984 | ||

Ispd
Genomic Map