siRNA Details
| Gene Symbol: Clec4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029465.3 | NP_083741.1 | 7 | 531 | ||
| GenBank mRNA | AK016282.1 | - | 1 | 36 | ||
| AK079992.1 | - | 7 | 532 | |||
| BC025069.1 | - | 7 | 523 | |||
| BC087963.1 | - | 7 | 531 | 6766377 | ||

Clec4g
Genomic Map