siRNA Details
| Gene Symbol: Vmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029478.3 | NP_083754.2 | 3 | 299 | ||
| GenBank mRNA | AB047550.1 | - | 3 | 311 | ||
| AB076609.1 | - | 2 | 190 | |||
| AK016312.1 | - | 3 | 316 | |||
| AK043091.1 | - | 3 | 300 | |||
| AK049559.1 | - | 3 | 292 | |||
| AK050796.1 | - | 2 | 168 | |||
| AK077443.1 | - | 3 | 299 | |||
| AK159468.1 | - | 3 | 314 | |||
| AK186168.1 | - | 2 | 218 | |||
| AX775963.1 | - | 3 | 269 | |||
| BC004013.1 | - | 3 | 293 | 3493738 | ||

Vmp1
Genomic Map