siRNA Details
| Gene Symbol: NDUFS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184979.1 | NP_001171908.1 | 3 | 362 | ||
| NM_004552.2 | NP_004543.1 | 3 | 366 | |||
| GenBank mRNA | AF020352.1 | - | 3 | 316 | ||
| AF047434.1 | - | 3 | 327 | |||
| AF086268.1 | - | 3 | 317 | |||
| AK312188.1 | - | 3 | 340 | |||
| BC001884.1 | - | 3 | 284 | 3544747 | ||
| BC093057.1 | - | 3 | 306 | 30382835 | ||
| CR456904.1 | - | 2 | 256 | |||

NDUFS5
Genomic Map