siRNA Details
| Gene Symbol: Srrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175229.3 | NP_780438.2 | 10 | 7136 | ||
| GenBank mRNA | AK090123.1 | - | 10 | 7097 | ||
| AK122248.1 | - | 1 | 4321 | |||
| BC026696.1 | - | 1 | 1954 | |||
| BC027781.1 | - | 1 | 1000 | |||

Srrm2
Genomic Map