siRNA Details
| Gene Symbol: Ccdc162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137750.1 | - | 4 | 497 | ||
| BC112401.1 | - | 10 | 1247 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc162| Gene Symbol: Ccdc162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137750.1 | - | 4 | 497 | ||
| BC112401.1 | - | 10 | 1247 | |||