siRNA Details
| Gene Symbol: Agmat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081408.1 | NP_001074877.1 | 5 | 968 | ||
| GenBank mRNA | AK017155.1 | - | 5 | 865 | ||
| BC025916.1 | - | 2 | 185 | |||
| BC037459.1 | - | 2 | 141 | |||
| BC115693.2 | - | 5 | 865 | 40104399 | ||

Agmat
Genomic Map