siRNA Details
| Gene Symbol: Zmym2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029498.3 | NP_083774.2 | 17 | 3066 | ||
| GenBank mRNA | AK137708.1 | - | 7 | 828 | ||
| AK137720.1 | - | 7 | 828 | |||
| AK164896.1 | - | 11 | 1293 | |||
| BC043450.1 | - | 14 | 1831 | |||
| BC084596.1 | - | 13 | 1673 | |||
| BC141403.1 | - | 17 | 3042 | 9056016 | ||

Zmym2
Genomic Map