siRNA Details
| Gene Symbol: Zc3h18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029993.1 | NP_001025164.1 | 8 | 1452 | ||
| NM_001029994.1 | NP_001025165.1 | 7 | 1380 | |||
| GenBank mRNA | AK031413.1 | - | 8 | 1401 | ||
| AK131189.1 | - | 5 | 524 | |||
| AK159881.1 | - | 8 | 1388 | |||
| AK165989.1 | - | 6 | 731 | |||
| BC030495.1 | - | 7 | 1360 | 5400494 | ||
| BC094330.1 | - | 5 | 533 | |||

Zc3h18
Genomic Map