siRNA Details
| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023799.3 | NP_076288.1 | 9 | 1623 | ||
| GenBank mRNA | AF132214.1 | - | 9 | 1468 | ||
| AK129188.1 | - | 9 | 1852 | |||
| AK145227.1 | - | 9 | 1873 | |||
| BC020093.1 | - | 9 | 1560 | |||
| BC041109.1 | - | 9 | 1953 | |||
| BC054821.1 | - | 9 | 1623 | 6515250 | ||

Mgea5
Genomic Map