siRNA Details
| Gene Symbol: Rnf183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153504.3 | NP_705724.1 | 3 | 870 | ||
| GenBank mRNA | AK017982.1 | - | 3 | 871 | ||
| BC025512.1 | - | 2 | 632 | 5252940 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf183| Gene Symbol: Rnf183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153504.3 | NP_705724.1 | 3 | 870 | ||
| GenBank mRNA | AK017982.1 | - | 3 | 871 | ||
| BC025512.1 | - | 2 | 632 | 5252940 | ||