siRNA Details
| Gene Symbol: Pcgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029508.3 | NP_083784.1 | 6, 7 | 745 | ||
| GenBank mRNA | AK002387.1 | - | 6, 7 | 768 | ||
| AK017984.1 | - | 6, 7 | 746 | |||
| AK046682.1 | - | 1, 2 | 503 | |||
| AK146133.1 | - | 6, 7 | 815 | |||
| BC139258.1 | - | 6, 7 | 706 | 8862280 | ||
| BC139259.1 | - | 6, 7 | 706 | 8862281 | ||
| BC145590.1 | - | 6, 7 | 706 | 9054156 | ||
| BC145591.1 | - | 6, 7 | 703 | 9054157 | ||

Pcgf5
Genomic Map