siRNA Details
| Gene Symbol: Pcgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029508.3 | NP_083784.1 | 8, 9 | 925 | ||
| GenBank mRNA | AK002387.1 | - | 8, 9 | 949 | ||
| AK017984.1 | - | 8, 9 | 926 | |||
| BC139258.1 | - | 8, 9 | 886 | 8862280 | ||
| BC139259.1 | - | 8, 9 | 886 | 8862281 | ||

Pcgf5
Genomic Map