siRNA Details
| Gene Symbol: Rap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029519.3 | NP_083795.2 | 2 | 734 | ||
| GenBank mRNA | AK075712.1 | - | 2 | 694 | ||
| AK140318.1 | - | 2 | 746 | |||
| BC043066.1 | - | 2 | 705 | 5709337 | ||
| BC049084.1 | - | 2 | 734 | 5706690 | ||
| BC053003.1 | - | 2 | 734 | 5706690 | ||
| BC067194.1 | - | 2 | 750 | |||

Rap2a
Genomic Map