siRNA Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029522.2 | NP_083798.2 | 12 | 1793 | ||
| GenBank mRNA | AK018084.1 | - | 2 | 40 | ||
| AK040996.1 | - | 12 | 1682 | |||
| AK143552.1 | - | 12 | 1794 | |||
| AY081187.1 | - | 12 | 1637 | |||
| BC021308.1 | - | 11 | 1348 | 5007832 | ||

Gpsm2
Genomic Map