siRNA Details
| Gene Symbol: Las1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152822.3 | NP_690035.2 | 1 | 288 | ||
| GenBank mRNA | AK018059.1 | - | 1 | 218 | ||
| AK089160.2 | - | 1 | 240 | |||
| AK163919.1 | - | 1 | 277 | |||
| BC032218.1 | - | 1 | 211 | |||
| BC141155.1 | - | 1 | 225 | 9055767 | ||
| BY036255.1 | - | 1 | 253 | |||
| BY059480.1 | - | 1 | 289 | |||

Las1l
Genomic Map