siRNA Details
| Gene Symbol: Depdc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172092.1 | NP_001165563.1 | 5 | 792 | ||
| NM_001172093.1 | NP_001165564.1 | 5 | 792 | |||
| NM_029523.3 | NP_083799.2 | 5 | 792 | |||
| GenBank mRNA | AK050456.1 | - | 5 | 729 | ||
| AK088719.1 | - | 5 | 768 | |||
| AK146458.1 | - | 5 | 761 | |||
| BC023934.1 | - | 5 | 785 | 5342167 | ||

Depdc1a
Genomic Map