siRNA Details
| Gene Symbol: Faxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175234.4 | NP_780443.2 | 3 | 760 | ||
| GenBank mRNA | AK018081.1 | - | 3 | 729 | ||
| AK082464.1 | - | 3 | 754 | |||
| AK144924.1 | - | 3 | 761 | |||
| BC059864.1 | - | 3 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Faxc| Gene Symbol: Faxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175234.4 | NP_780443.2 | 3 | 760 | ||
| GenBank mRNA | AK018081.1 | - | 3 | 729 | ||
| AK082464.1 | - | 3 | 754 | |||
| AK144924.1 | - | 3 | 761 | |||
| BC059864.1 | - | 3 | 274 | |||